APO455827436 Apollo Scientific Ltd

3-(Bromomethyl)benzaldehyde

  • MDL number MFCD06797497
  • Cas Number : 82072-23-9
  • Molecular Weight : 199.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 49-51
Iupac Name 3-(bromomethyl)benzaldehyde
Inchi Code 1S/C8H7BrO/c9-5-7-2-1-3-8(4-7)6-10/h1-4,6H,5H2
Inchi Key OEPGAYXSRGROSQ-UHFFFAOYSA-N

Description

Synonym 3-Formylbenzyl bromide
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 49-51
Product Link https://store.apolloscientific.co.uk/product/3-bromomethylbenzaldehyde

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromomethylbenzaldehyde
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart