APO455827451 Apollo Scientific Ltd

2-Nitro-L-phenylalanine

  • MDL number MFCD00270360
  • Cas Number : 19883-75-1
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 226-227
Iupac Name (2S)-2-amino-3-(2-nitrophenyl)propanoic acid
Inchi Code 1S/C9H10N2O4/c10-7(9(12)13)5-6-3-1-2-4-8(6)11(14)15/h1-4,7H,5,10H2,(H,12,13)/t7-/m0/s1
Inchi Key SDZGVFSSLGTJAJ-ZETCQYMHSA-N

Description

Synonym (2S)-2-Amino-3-(2-nitrophenyl)propanoic acid, (2S)-2-Amino-3-(2-nitrophenyl)propionic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 226-227
Product Link https://store.apolloscientific.co.uk/product/2-nitro-l-phenylalanine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-nitro-l-phenylalanine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart