APO455827460 Apollo Scientific Ltd

3-(Benzo[b]thiophen-3-yl)-L-alanine

  • MDL number MFCD00079684
  • Cas Number : 72120-71-9
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 248(dec.)
Iupac Name (2S)-2-amino-3-(1-benzothien-3-yl)propanoic acid
Inchi Code 1S/C11H11NO2S/c12-9(11(13)14)5-7-6-15-10-4-2-1-3-8(7)10/h1-4,6,9H,5,12H2,(H,13,14)/t9-/m0/s1
Inchi Key GAUUPDQWKHTCAX-VIFPVBQESA-N

Description

Synonym 3-(1-Benzothiophen-3-yl)-L-alanine, 3-(Thianaphthen-3-yl)-L-alanine, (2S)-2-Amino-3-(benzo[b]thiophen-3-yl)propanoic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 248(dec.)
Product Link https://store.apolloscientific.co.uk/product/3-benzobthiophen-3-yl-l-alanine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-benzobthiophen-3-yl-l-alanine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart