APO455827461 Apollo Scientific Ltd

2-Allyl-L-glycine

  • MDL number MFCD00002627
  • Cas Number : 16338-48-0
  • Molecular Weight : 115.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 283(dec.)
Iupac Name (2S)-2-amino-4-pentenoic acid
Inchi Code 1S/C5H9NO2/c1-2-3-4(6)5(7)8/h2,4H,1,3,6H2,(H,7,8)/t4-/m0/s1
Inchi Key WNNNWFKQCKFSDK-BYPYZUCNSA-N

Description

Synonym (2S)-2-Aminopent-4-enoic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 283(dec.)
Product Link https://store.apolloscientific.co.uk/product/2-allyl-l-glycine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-allyl-l-glycine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart