APO455827463 Apollo Scientific Ltd

1-(3-Chloropropyl)-4-methylpiperazine dihydrochloride

  • MDL number MFCD00023304
  • Cas Number : 2031-23-4
  • Molecular Weight : 249.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-chloropropyl-4-methylpiperazine-dihydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-chloropropyl-4-methylpiperazine-dihydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart