APO455827467 Apollo Scientific Ltd

3-(Aminomethyl)phenol

  • MDL number MFCD00798977
  • Cas Number : 73604-31-6
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-168
Synonym 3-Hydroxybenzylamine, (3-Hydroxyphenyl)methylamine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-aminomethylphenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminomethylphenol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart