APO455827468 Apollo Scientific Ltd

2-Hydroxy-4-methoxy-5-nitropyridine

  • MDL number MFCD07367779
  • Cas Number : 607373-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Methoxy-5-nitropyridin-2-ol
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40784

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40784
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart