APO455827472 Apollo Scientific Ltd

5-Amino-2,4-dichloro-6-methylpyrimidine

  • MDL number MFCD08062586
  • Cas Number : 13162-27-1
  • Molecular Weight : 178.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichloro-6-methyl-5-pyrimidinamine
Inchi Code 1S/C5H5Cl2N3/c1-2-3(8)4(6)10-5(7)9-2/h8H2,1H3
Inchi Key FDMMHWIFYCXEOF-UHFFFAOYSA-N

Description

Synonym 2,4-Dichloro-6-methylpyrimidin-5-amine, 5-Amino-2,4-dichloro-6-methyl-1,3-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-24-dichloro-6-methylpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-24-dichloro-6-methylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart