APO455827473 Apollo Scientific Ltd

Ethyl 2-amino-1,3-thiazole-4-carboxylate hydrobromide

  • MDL number MFCD06653384
  • Cas Number : 127942-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-177
Synonym 2-Amino-4-(ethoxycarbonyl)-1,3-thiazole hydrobromide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-thiazole-4-carboxylate-hydrobromide

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-thiazole-4-carboxylate-hydrobromide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart