APO455827479 Apollo Scientific Ltd

2-(4-Aminophenyl)propanenitrile

  • MDL number MFCD09865026
  • Cas Number : 28694-90-8
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-57
Synonym (4-Aminophenyl)(methyl)acetonitrile, 4-(1-Cyanoethyl)aniline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-aminophenylpropanenitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-aminophenylpropanenitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart