APO455827481 Apollo Scientific Ltd

1-(2-Methoxyphenyl)-3-methylpiperazine

  • MDL number MFCD06804057
  • Cas Number : 152943-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3-Methylpiperazin-1-yl)anisole, 1-Methoxy-2-(3-methylpiperazin-1-yl)benzene
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41242

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41242
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart