APO455827489 Apollo Scientific Ltd

4-Isocyanato-4-(thien-2-yl)tetrahydro-2H-pyran

  • MDL number MFCD09879931
  • Cas Number : 926921-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49.5-52.5
Synonym 4-(Thien-2-yl)tetrahydro-2H-pyran-4-yl isocyanate, 2-(4-Isocyanatotetrahydro-2H-pyran-4-yl)thiophene
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41498

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41498
pictograms GHS07;GHS08
hstatements H302+H312+H332;H334
Availability details for
Quantity
  • Availability
  • Add to Cart