APO455827503 Apollo Scientific Ltd

[2-(Thien-2-yl)pyrimidin-5-yl]methanol

  • MDL number MFCD09863233
  • Cas Number : 921939-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115.5-118
Synonym 5-(Hydroxymethyl)-2-(thien-2-yl)pyrimidine, 2-[5-(Hydroxymethyl)pyrimidin-2-yl]thiophene
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41558

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41558
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart