APO455827511 Apollo Scientific Ltd

3-(Aminomethyl)-5-bromopyridine

  • MDL number MFCD06212876
  • Cas Number : 135124-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromopyridin-3-yl)methylamine, alpha-Amino-5-bromo-3-picoline
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42202

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42202
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart