APO455827512 Apollo Scientific Ltd

4-(3-Bromothien-2-yl)phenacyl bromide

  • MDL number MFCD09879969
  • Cas Number : 944450-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-47.5
Synonym 2-Bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethan-1-one
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42211

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42211
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart