APO455827516 Apollo Scientific Ltd

2-Amino-4-nitrobenzonitrile

  • MDL number MFCD04117876
  • Cas Number : 87376-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-5-nitroaniline, 3-Amino-4-cyanonitrobenzene
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-nitrobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-nitrobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart