APO455827520 Apollo Scientific Ltd

4-(3,5-Dimethyl-1H-pyrazol-1-yl)benzaldehyde

  • MDL number MFCD06800625
  • Cas Number : 934570-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Synonym 3,5-Dimethyl-1-(4-formylphenyl)-1H-pyrazole
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42234

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42234
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart