APO455827523 Apollo Scientific Ltd

3,4,5-Trichloropyridazine

  • MDL number MFCD00006464
  • Cas Number : 14161-11-6
  • Molecular Weight : 183.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 54-61
Iupac Name 3,4,5-trichloropyridazine
Inchi Code 1S/C4HCl3N2/c5-2-1-8-9-4(7)3(2)6/h1H
Inchi Key GBAOOJAWDCNOGO-UHFFFAOYSA-N

Description

Synonym 3,4,5-Trichloro-1,2-diazine
Purity 99%
Flash Point 117-118/14mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 54-61
Product Link https://store.apolloscientific.co.uk/product/345-trichloropyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/345-trichloropyridazine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart