APO455827528 Apollo Scientific Ltd

3-Amino-6-(4-methoxyphenyl)pyridazine

  • MDL number MFCD06801353
  • Cas Number : 4776-87-8
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(4-methoxyphenyl)-3-pyridazinamine
Inchi Code 1S/C11H11N3O/c1-15-9-4-2-8(3-5-9)10-6-7-11(12)14-13-10/h2-7H,1H3,(H2,12,14)
Inchi Key PCPMUYYPWJFHEQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-4-methoxyphenylpyridazine

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-4-methoxyphenylpyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart