APO455827533 Apollo Scientific Ltd

2-Amino-4-nitrobenzyl alcohol

  • MDL number MFCD01664410
  • Cas Number : 78468-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Amino-4-nitrophenyl)methanol, 2-(Hydroxymethyl)-5-nitroaniline
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-nitrobenzyl-alcohol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart