APO455827541 Apollo Scientific Ltd

2,5-Dichloroanisole

  • MDL number MFCD00061123
  • Cas Number : 1984-58-3
  • Molecular Weight : 177.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name 1,4-dichloro-2-methoxybenzene
density 1.33
Inchi Code 1S/C7H6Cl2O/c1-10-7-4-5(8)2-3-6(7)9/h2-4H,1H3
Inchi Key QKMNFFSBZRGHDJ-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 1,4-Dichloro-2-methoxybenzene, 2,5-Dichlorophenyl methyl ether
Purity 98%
Density 1.33
Flash Point 128-132/3.5mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-dichloroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-dichloroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart