APO455827546 Apollo Scientific Ltd

5-Bromopyridine-2-carbonyl chloride

  • MDL number MFCD07368290
  • Cas Number : 137178-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120(dec.)
Synonym 5-Bromopicolinoyl chloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromopyridine-2-carbonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromopyridine-2-carbonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart