APO455827547 Apollo Scientific Ltd

2,3-Dimethylchlorobenzene

  • MDL number MFCD00060689
  • Cas Number : 608-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2,3-dimethylbenzene, 3-Chloro-o-xylene
Purity 98
Density 1.1
Flash Point 73
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 194
Product Link https://store.apolloscientific.co.uk/product/23-dimethylchlorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-dimethylchlorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart