APO455827550 Apollo Scientific Ltd

Methyl 6-methoxynicotinate

  • MDL number MFCD00674130
  • Cas Number : 26218-80-4
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 48-52
Iupac Name methyl 6-methoxynicotinate
Inchi Code 1S/C8H9NO3/c1-11-7-4-3-6(5-9-7)8(10)12-2/h3-5H,1-2H3
Inchi Key OVLDWZNVBDRZNN-UHFFFAOYSA-N

Description

Synonym Methyl 6-methoxypyridine-3-carboxylate, 2-Methoxy-5-(methoxycarbonyl)pyridine
Flash Point 92/3.5mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 48-52
Product Link https://store.apolloscientific.co.uk/product/methyl-6-methoxynicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-methoxynicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart