APO455827560 Apollo Scientific Ltd

3-Chloro-5-iodobenzonitrile

  • MDL number MFCD00672974
  • Cas Number : 289039-30-1
  • Molecular Weight : 263.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-iodobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-iodobenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart