APO455827561 Apollo Scientific Ltd

Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)nicotinate

  • MDL number MFCD01311995
  • Cas Number : 402479-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 5-cyano-6-methyl-2-(piperazin-1-yl)pyridine-3-carboxylate, 3-Cyano-5-(ethoxycarbonyl)-2-methyl-6-(piperazin-1-yl)pyridine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29017

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29017
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart