APO455827562 Apollo Scientific Ltd

4-(Piperazin-1-yl)thieno[3,2-d]pyrimidine

  • MDL number MFCD03407481
  • Cas Number : 683274-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Thieno[3,2-d]pyrimidin-4-yl)piperazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29019

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29019
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart