APO455827567 Apollo Scientific Ltd

5-Chloro-2-(1H-tetrazol-5-yl)aniline

  • MDL number MFCD03085917
  • Cas Number : 54013-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Synonym 5-(2-Amino-4-chlorophenyl)-1H-tetrazole
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29030

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29030
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart