APO455827572 Apollo Scientific Ltd

4-(Chloromethyl)piperidine, N-BOC protected

  • MDL number MFCD08689676
  • Cas Number : 479057-79-9
  • Molecular Weight : 233.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 49-51
Iupac Name tert-butyl 4-(chloromethyl)-1-piperidinecarboxylate
Inchi Code 1S/C11H20ClNO2/c1-11(2,3)15-10(14)13-6-4-9(8-12)5-7-13/h9H,4-8H2,1-3H3
Inchi Key CTSABEXZMPJSGO-UHFFFAOYSA-N

Description

Synonym tert-Butyl 4-(chloromethyl)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(chloromethyl)piperidine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 49-51
Product Link https://store.apolloscientific.co.uk/product/4-chloromethylpiperidine-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloromethylpiperidine-n-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart