APO455827583 Apollo Scientific Ltd

4-(Phthalimid-1-yl)butanoic acid

  • MDL number MFCD00196079
  • Cas Number : 3130-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-121
Synonym 4-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)butanoic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-phthalimid-1-ylbutanoic-acid

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-phthalimid-1-ylbutanoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart