APO455827588 Apollo Scientific Ltd

Ethyl 4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD08444006
  • Cas Number : 924869-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149
Synonym 5-(Ethoxycarbonyl)-4-hydroxy-2-(4-methoxyphenyl)-1,3-thiazole, 4-[5-(Ethoxycarbonyl)-4-hydroxy-1,3-thiazol-2-yl]anisole
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39040

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39040
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart