APO455827595 Apollo Scientific Ltd

3-Oxo-3-pyridin-3-ylpropanenitrile

  • MDL number MFCD08282797
  • Cas Number : 30510-18-0
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 79-81
Iupac Name 3-oxo-3-(3-pyridinyl)propanenitrile
Inchi Code 1S/C8H6N2O/c9-4-3-8(11)7-2-1-5-10-6-7/h1-2,5-6H,3H2
Inchi Key MDNYQXMCDAKSIW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 79-81
Product Link https://store.apolloscientific.co.uk/product/3-oxo-3-pyridin-3-ylpropanenitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-oxo-3-pyridin-3-ylpropanenitrile
pictograms GHS06;GHS07
hstatements H315;H319;H330;H335
Availability details for
Quantity
  • Availability
  • Add to Cart