APO455827596 Apollo Scientific Ltd

3-Oxo-3-(pyridin-2-yl)propanenitrile

  • MDL number MFCD06656806
  • Cas Number : 54123-21-6
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-99
Synonym 2-(Cyanoacetyl)pyridine, 2-(2-Cyanoethanoyl)pyridine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-oxo-3-pyridin-2-ylpropanenitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-oxo-3-pyridin-2-ylpropanenitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart