APO455827599 Apollo Scientific Ltd

2-Amino-4-hydroxy-5-phenyl-1,3-thiazole

  • MDL number MFCD08444003
  • Cas Number : 98879-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-242
Synonym 2-Amino-5-phenyl-1,3-thiazol-4-ol, 4-Hydroxy-5-phenyl-1,3-thiazol-2-amine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39116

Safety Information

signalword Warning
pcodes P280;P260
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39116
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart