APO455827609 Apollo Scientific Ltd

2-Chloro-6-iodonitrobenzene

  • MDL number MFCD08443947
  • Cas Number : 937601-42-8
  • Molecular Weight : 283.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 60-61
Iupac Name 1-chloro-3-iodo-2-nitrobenzene
Inchi Code 1S/C6H3ClINO2/c7-4-2-1-3-5(8)6(4)9(10)11/h1-3H
Inchi Key WRRLPIZQDPPPJS-UHFFFAOYSA-N

Description

Synonym 1-Chloro-3-iodo-2-nitrobenzene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-61
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-iodonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-iodonitrobenzene
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart