APO455827611 Apollo Scientific Ltd

2-(4-Chlorophenyl)-4-hydroxy-5-phenyl-1,3-thiazole

  • MDL number MFCD08444002
  • Cas Number : 65752-50-3
  • Molecular Weight : 287.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252-254
Synonym 2-(4-Chlorophenyl)-5-phenyl-1,3-thiazol-4-ol
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-chlorophenyl-4-hydroxy-5-phenyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-4-chlorophenyl-4-hydroxy-5-phenyl-13-thiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart