APO455827616 Apollo Scientific Ltd

(2E)-3-(Dimethylamino)-1-phenylprop-2-en-1-one

  • MDL number MFCD00121193
  • Cas Number : 1201-93-0
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 91-93
Iupac Name (2E)-3-(dimethylamino)-1-phenyl-2-propen-1-one
Inchi Code 1S/C11H13NO/c1-12(2)9-8-11(13)10-6-4-3-5-7-10/h3-9H,1-2H3/b9-8+
Inchi Key HUTKDPINCSJXAA-CMDGGOBGSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91-93
Product Link https://store.apolloscientific.co.uk/product/2e-3-dimethylamino-1-phenylprop-2-en-1-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2e-3-dimethylamino-1-phenylprop-2-en-1-one
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart