APO455827619 Apollo Scientific Ltd

Ethyl 2,4-dioxo-4-(2-hydroxy-5-methyl-3-nitrophenyl)butanoate

  • MDL number MFCD08443987
  • Cas Number : 859843-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Synonym Ethyl 2,4-dioxo-4-(2-hydroxy-5-methyl-3-nitrophenyl)butyrate, 2-(3,4-Dioxo-4-ethoxybutanoyl)-4-methyl-6-nitrophenol
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39227

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39227
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart