APO455827624 Apollo Scientific Ltd

5-Hydroxyquinolin-2(1H)-one

  • MDL number MFCD02181132
  • Cas Number : 31570-97-5
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp >300
Iupac Name 2,5-quinolinediol
Inchi Code 1S/C9H7NO2/c11-8-3-1-2-7-6(8)4-5-9(12)10-7/h1-5,11H,(H,10,12)
Inchi Key XOXGLLQTNQBDKL-UHFFFAOYSA-N

Description

Synonym 1,2-Dihydro-5-hydroxy-2-oxoquinoline
Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point >300
Product Link https://store.apolloscientific.co.uk/product/5-hydroxyquinolin-21h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxyquinolin-21h-one
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart