APO455827625 Apollo Scientific Ltd

3-Methoxybenzoylacetonitrile

  • MDL number MFCD01566517
  • Cas Number : 21667-60-7
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Synonym 3-(3-Methoxyphenyl)-3-oxopropanenitrile, 3-(3-Methoxyphenyl)-3-oxopropionitrile
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methoxybenzoylacetonitrile

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/3-methoxybenzoylacetonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart