APO455827626 Apollo Scientific Ltd

Methyl 4-amino-3-nitrobenzoate

  • MDL number MFCD00017562
  • Cas Number : 3987-92-6
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-200
Synonym 2-Amino-5-(methoxycarbonyl)nitrobenzene, 4-(Methoxycarbonyl)-2-nitroaniline
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-amino-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-amino-3-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart