APO455827629 Apollo Scientific Ltd

Methyl 4-amino-3-bromo-5-nitrobenzoate

  • MDL number MFCD08443938
  • Cas Number : 105655-17-2
  • Molecular Weight : 275.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 108-110
Iupac Name methyl 4-amino-3-bromo-5-nitrobenzoate
Inchi Code 1S/C8H7BrN2O4/c1-15-8(12)4-2-5(9)7(10)6(3-4)11(13)14/h2-3H,10H2,1H3
Inchi Key PRTLMEWWYBJZPN-UHFFFAOYSA-N

Description

Synonym 2-Bromo-4-(methoxycarbonyl)-6-nitroaniline, 2-Amino-3-bromo-5-(methoxycarbonyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 108-110
Product Link https://store.apolloscientific.co.uk/product/methyl-4-amino-3-bromo-5-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-amino-3-bromo-5-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart