APO455827634 Apollo Scientific Ltd

4-(2-Chloroethyl)piperazine, N1-BOC protected

  • MDL number MFCD08443993
  • Cas Number : 208167-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Synonym tert-Butyl 4-(2-chloroethyl)piperazine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(2-chloroethyl)piperazine
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39385

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39385
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart