APO455827640 Apollo Scientific Ltd

3,6-Dichloro[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD00139673
  • Cas Number : 33050-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-143
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/36-dichloro124triazolo43-bpyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/36-dichloro124triazolo43-bpyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart