APO455827648 Apollo Scientific Ltd

5'-Chloro-2'-nitroacetophenone

  • MDL number MFCD03787322
  • Cas Number : 18640-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-61
Synonym 1-(5-Chloro-2-nitrophenyl)ethan-1-one
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39470

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39470
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart