APO455827650 Apollo Scientific Ltd

Ethyl 2-aminonicotinate

  • MDL number MFCD01569452
  • Cas Number : 13362-26-0
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 96-98
Iupac Name ethyl 2-aminonicotinate
Inchi Code 1S/C8H10N2O2/c1-2-12-8(11)6-4-3-5-10-7(6)9/h3-5H,2H2,1H3,(H2,9,10)
Inchi Key KIAGEDYOPMHRRB-UHFFFAOYSA-N

Description

Synonym Ethyl 2-aminopyridine-3-carboxylate
Purity 98%
Flash Point 266.75
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 96-98
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-aminonicotinate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-aminonicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart