APO455827652 Apollo Scientific Ltd

4-(4-Ethylpiperazin-1-yl)aniline

  • MDL number MFCD00702246
  • Cas Number : 115619-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-78
Synonym 1-(4-Aminophenyl)-4-ethylpiperazine
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-4-ethylpiperazin-1-ylaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-ethylpiperazin-1-ylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart