APO455827660 Apollo Scientific Ltd

4-Amino-1,3-benzoxazole

  • MDL number MFCD00973373
  • Cas Number : 163808-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-79
Synonym 1,3-Benzoxazol-4-amine, 4-Aminobenzo[d]oxazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-13-benzoxazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-13-benzoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart