APO455827661 Apollo Scientific Ltd

1-[2-Amino-5-(morpholin-4-yl)phenyl]ethanone

  • MDL number MFCD03787743
  • Cas Number : 98440-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-105
Synonym 2'-Amino-5'-(morpholin-4-yl)acetophenone, 2-Acetyl-4-(morpholin-4-yl)aniline
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39569

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39569
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart