APO455827663 Apollo Scientific Ltd

4-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD07838483
  • Cas Number : 875160-43-3
  • Molecular Weight : 204.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Synonym 4-Bromo-2-carboxy-1-methyl-1H-pyrrole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-pyrrole-2-carboxylic-acid

Safety Information

MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-pyrrole-2-carboxylic-acid
Availability details for
Quantity
  • Availability
  • Add to Cart